Learn More
Kuhner Shaker Inc STERILE SINGLE-USE BAG
Supplier:  Kuhner Shaker Inc NSMX760004
Encompass
Pack of 3 Standard Sterile Single-Use Bags for use with Kuhner Shaker's SB10-X Orbitally Shaken Bioreactors with 3L Vessel Adapter. Includes bio-weldable tubing and integrated (non-invasive) pH and DO Optical Sensors. (H)
Catalog No. 50-257-0321