Learn More
Andwin Scientific STERILE SLEEVE XL 300/CS
SDP
Catalog No. NC0913648
Encompass_Preferred
The Sterile Sleeves are made from specially formulated breathable microporous fabric that gives users great fluid and barrier protection. This product also features durability and elasticity. The sleeves are gamma irradiated to a Sterility Assurance
This item is not returnable. View return policy
Catalog No. NC0913648 Supplier Andwin Scientific Supplier No. 414004511
The Sterile Sleeves are made from specially formulated breathable microporous fabric that gives users great fluid and barrier protection. This product also features durability and elasticity. The sleeves are gamma irradiated to a Sterility Assurance