Learn More
Benchmark Products Inc STERILE SUITE-COVER ISOLATERS
SDP
Catalog No. NC0190197
Encompass
Sterile cover for the Suite�-Reach tool: a perfect for cleaning and applying disinfectants in isolators, biological safety cabinets, glove boxes and other hard to reach spaces. The custom cover made out of the revolutionary bi-component ultra-fiber Suite� material technology, this is the perfect device for both cleaning and applying. The Suite�-Reach tool is lightweight, extends from 21"-37", swivels 3600 and is fully autoclavable.
This item is not returnable. View return policy
Catalog No. NC0190197 Supplier Benchmark Products Inc Supplier No. YTTCIBDI20S
Sterile cover for the Suite�-Reach tool: a perfect for cleaning and applying disinfectants in isolators, biological safety cabinets, glove boxes and other hard to reach spaces. The custom cover made out of the revolutionary bi-component ultra-fiber Suite� material technology, this is the perfect device for both cleaning and applying. The Suite�-Reach tool is lightweight, extends from 21"-37", swivels 3600 and is fully autoclavable.