Learn More
Haztech Systems, Inc HW4089 Sterile single tipped applicators individually wrapped swabs - 10 Swabs included
SDP
Catalog No. NC9753307
Encompass
HW4089 Sterile Polyester Tipped Swabs with 6-Inch Semi-Flexible Plastic Shaft & 0.20 x 0.69-Inch polyester tip, Individually Wrapped - 10 individually wrapped Swabs included. Uses include collection of biological specimens for DNA testing, collecting biological samples from surfaces.
Catalog No. NC9753307 Supplier Haztech Systems, Inc Supplier No. HW4089
HW4089 Sterile Polyester Tipped Swabs with 6-Inch Semi-Flexible Plastic Shaft & 0.20 x 0.69-Inch polyester tip, Individually Wrapped - 10 individually wrapped Swabs included. Uses include collection of biological specimens for DNA testing, collecting biological samples from surfaces.