Learn More
Research Products International Corp Sterile Syringe Filters, Disposable, Nylon Membrane, .45m Pore Size, 30mm Diameter, 30 per Pack
SDP
Supplier:  Research Products International Corp 256133
Encompass
Syringe filters are ideal for filtering culture media and organic solvents in a wide range of applications. All items are tested for filter efficiency and housing integrity. Filter housing is manufactured and tested to withstand pressures of up to 75 PSI (5.1 Bar).
Mainly used for pre-filtration and purification to sterilize and make samples of solution particle-free and sterile. Designed for low hold-up to minimize sample loss. Lot numbers are printed on sterile packs for easy traceability. Sterile filters are packaged in a convenient, peel-open package. All products come certified non-pyrogenic and Gamma irradiated. RNase and DNase free.
Catalog No. 50-476-575