Learn More
Fisherbrand™ Sterile Plastic Syringe, Laboratory Use Only
Catalog No. 14955570 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 1 mL
10 mL
20 mL
3 mL
5 mL
60 mL
Connection Type:
Luer Lock
Luer Slip
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Connection Type
14-955-570 20 mL Luer Slip
14-955-560 0.1 to 1 mL Luer Lock
14-955-561 3 mL Luer Lock
14-955-562 5 mL Luer Lock
14-955-563 10 mL Luer Lock
14-955-564 20 mL Luer Lock
14-955-565 60 mL Luer Lock
14-955-566 0.1 to 1 mL Luer Slip
14-955-567 3 mL Luer Slip
14-955-568 5 mL Luer Slip
14-955-569 10 mL Luer Slip
14-955-571 60 mL Luer Slip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14-955-570
Add to Cart
Edge
Add to Cart
Syringes provide accurate measurements and are available in a wide range of sizes.
  • High-transparency barrel to easily visualize contents
  • Silicone lubrication ensures smooth plunger movement
  • Plunger backstop that prevents accidental withdrawal of plunger from the barrel
  • Large finger grips allow for easy, controlled injections
Specifications
Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge No Needle Included
For Use With (Application) Laboratory use only, not intended for medication dispensing
Includes Latex free rubber plunger with silicone oil lubrication
Latex Free Yes
Product Type Syringe
Volume (Metric) 20 mL
Connection Type Luer Slip
Graduated Yes
Sterility Sterile