Learn More
Mascon Medical Mascon Medical™ Sterile Syringe Filter, Sterile, 0.22μm*30mm, 50EA/Box, 10 Box/Case
SDP
Supplier:  Mascon Medical FSNEEDLEFILTER
Encompass_Preferred
Our Mascon Medical™ high performance Syringe Filters are made of medical-grade polypropylene making them a perfect match for disposable syringes. The syringe filters are mainly used for filtering small volume samples, such as protein, buffer and drug samples. Features:•Meets ISO13485 quality management system standard•RNase/DNase free•Non-pyrogenic•Endotoxin-free•Adaptation: injection syringeSpecifications:•Volume: 0.22um*30mm•Qty. / Case: 500•Material: Polypropylene, PES filter membrane•Sterile: Yes•Size: Φ33.6*23.4mm•Color: Green•Temperature Range: -40℃~100℃•Packaging Format: 1 EA/Bag, 50 Bags/Box, 10 Box/Case
Catalog No. NC2458333