Learn More
Sony Biotechnology Sterile Tubing (Process Development)
Catalog No. NC3885083
Encompass_Preferred
Consumable, single-use kit for use with the CGX10 Cell Isolation System - 3 kits/box, where each kit includes:
• Sterile Tubing (Process Development) (1)
• Sterile Tubing Extender (Buffer Bag) (1)
• Sterile Tubing Extender (Sheath Bag) (1)
Catalog No. NC3885083 Supplier Sony Biotechnology Supplier No. LECGXTP1
Add to Cart
Add to Cart
Request a Quote
Consumable, single-use kit for use with the CGX10 Cell Isolation System - 3 kits/box, where each kit includes:
• Sterile Tubing (Process Development) (1)
• Sterile Tubing Extender (Buffer Bag) (1)
• Sterile Tubing Extender (Sheath Bag) (1)