Learn More
Sony Biotechnology Sterile Tubing (Standard Operation)
Catalog No. NC3885086
Encompass_Preferred
Consumable, single-use kit for use with the CGX10 Cell Isolation System - 3 kits/box, where each kit includes:
• Sterile Tubing (Standard Operation) (1)
• Sterile Tubing Extender (Buffer Bag) (1)
• Sterile Tubing Extender (Sheath Bag) (1)
• Sterile Sample Preparation Bag (1)
Catalog No. NC3885086 Supplier Sony Biotechnology Supplier No. LECGXTS1
Add to Cart
Add to Cart
Request a Quote
Consumable, single-use kit for use with the CGX10 Cell Isolation System - 3 kits/box, where each kit includes:
• Sterile Tubing (Standard Operation) (1)
• Sterile Tubing Extender (Buffer Bag) (1)
• Sterile Tubing Extender (Sheath Bag) (1)
• Sterile Sample Preparation Bag (1)