Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Sterile Universal Filter Tips

Catalog No. 22171783
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 100 μL
1 to 1000 μL
1 to 20 μL
1 to 200 μL
1 to 300 μL
Length (Metric):
32 mm
50 mm
59 mm
86 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Length (Metric)
22-171-783 1 to 1000 μL 86 mm
22-171-777 0.1 to 10 μL 32 mm
22-171-779 1 to 20 μL 50 mm
22-171-780 1 to 100 μL 50 mm
22-171-781 1 to 200 μL 59 mm
22-171-782 1 to 300 μL 59 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-171-783 Supplier Globe Scientific Supplier No. 150935
Only null left
Add to Cart
Add to Cart

Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems.

  • Manufactured from clear polypropylene (PP)
  • Polypropylene (PP) filters protect samples and pipettes from contamination
  • Dnase-free, Rnase-free and non-pyrogenic
  • Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems
  • Sterilized by gamma irradiation SAL10-6 (ISO11137)
  • Racks are printed with lot number for quality traceability

Specifications

Volume (Metric) 1 to 1000 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Length (Metric) 86 mm
Clean Claims Rnase, Dnase, and Pyrogen Free
Sterilization Method Gamma Irradiation
Format Rack
Tip Style Standard
Packaging 96 Tips/Rack, 10 Racks/Pk, 2 Boxes/Cs.
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.