Learn More
Terumo BCT Inc TSCD/TSCD-II/TSCD-Q Wafers
Catalog No. NC0341972
Encompass_Preferred
The wafers used by the TSCD-II welder heat up to 300 ̊C, maintaining sterility during cutting and welding without allowing particulates or chemical residue to form in the tubes.
This item is not returnable. View return policy
Catalog No. NC0341972 Supplier Terumo BCT Inc Supplier No. 1SCW017
The wafers used by the TSCD-II welder heat up to 300 ̊C, maintaining sterility during cutting and welding without allowing particulates or chemical residue to form in the tubes.