Learn More
Hanna Pharmaceutical Supply Co., Inc. STERILE WATER F/INJCTN 100ML
Catalog No. NC2468381
Encompass_Preferred
  • Sterile and non-pyrogenic solution for diluting or dissolving drugs
  • Single-dose glass fliptop vial ensures purity and integrity
  • Hypotonic solution that requires mixing with appropriate solutes to be isotonic
  • Preservative-free for safe preparation of medications
  • Not for direct intravenous administration
  • Stored in a temperature-controlled environment of 20 to 25°C (68 to 77°F)
  • 25 vials packaged per tray/case
  • Not made with natural rubber latex and non-DEHP
This item is not returnable. View return policy
Catalog No. NC2468381 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. 00409488799
  • Sterile and non-pyrogenic solution for diluting or dissolving drugs
  • Single-dose glass fliptop vial ensures purity and integrity
  • Hypotonic solution that requires mixing with appropriate solutes to be isotonic
  • Preservative-free for safe preparation of medications
  • Not for direct intravenous administration
  • Stored in a temperature-controlled environment of 20 to 25°C (68 to 77°F)
  • 25 vials packaged per tray/case
  • Not made with natural rubber latex and non-DEHP