Learn More
Lakeside Manufacturing Inc Sterile Wrap Rack with Hang Bars, 1 Shelf: 18" x 60"
Catalog No. 502884454
Encompass_Preferred
Sterile Wrap Racks by Lakeside Manufacturing have adjustable hang bars that keep sterile wraps separated from each other and easily accessible. The Lakeside R1860SWR Sterile Wrap Rack with hang bars has (1) shelf measuring 18" x 60". This Lakeside rack's bottom wire shelf allows for additional product to be stored and readily available. Its 5" (127) casters provide smooth maneuverability around tight spaces. This rack's chrome plated wire shelf and posts are easy to clean and sanitize. Its (2) horizontal hang bars hold and sterilize medical wrap. Its hang bars are adjustable to keep sterile wraps separated from each other and easily accessible. Endless variations and feature combinations. Superior lead times. Made in the USA.
Catalog No. 50-288-4454 Supplier Lakeside Manufacturing Inc Supplier No. R1860SWR
Sterile Wrap Racks by Lakeside Manufacturing have adjustable hang bars that keep sterile wraps separated from each other and easily accessible. The Lakeside R1860SWR Sterile Wrap Rack with hang bars has (1) shelf measuring 18" x 60". This Lakeside rack's bottom wire shelf allows for additional product to be stored and readily available. Its 5" (127) casters provide smooth maneuverability around tight spaces. This rack's chrome plated wire shelf and posts are easy to clean and sanitize. Its (2) horizontal hang bars hold and sterilize medical wrap. Its hang bars are adjustable to keep sterile wraps separated from each other and easily accessible. Endless variations and feature combinations. Superior lead times. Made in the USA.