Learn More
Research Products International Corp Sterile Cloning Discs, 6.0mm (1/4 Inch) Diameter, 100 per Package
SDP
Catalog No. 50550300
Encompass
Convenient method for isolating a large number of clones. Sterile paper discs allow transfer of cells to a multi-well plate in a fraction of the time normally required and reduce possibility of contamination. Discs are sterilized by Gamma radiation. Supplied in two sterile tubes each containing fifty discs.
Catalog No. 50-550-300 Supplier Research Products International Corp Supplier No. 248712
Convenient method for isolating a large number of clones. Sterile paper discs allow transfer of cells to a multi-well plate in a fraction of the time normally required and reduce possibility of contamination. Discs are sterilized by Gamma radiation. Supplied in two sterile tubes each containing fifty discs.