Learn More
Avantor Fluid Handling SterilEnz-II/AT, 1/2"""" HB x Mini Flange Adapter, Polypropylene, Silicone Gasket, Autoclaveable, 10/pk
Catalog No. NC1767791
Encompass
Standard Sanitary Fittings for Single-Use Systems. SterilEnz-II/AT connectors come with a platinum cured, medical grade silicone gasket mechanically attached to the fitting face. SterilEnz-II/AT fittings are made rugged inert polypropylene suitable for either autoclaving or gamma-irradiation. The new II/AT series is available in seven fitting sizes.
This item is not returnable. View return policy
Catalog No. NC1767791 Supplier Avantor Fluid Handling Supplier No. 2AT812TC10
Standard Sanitary Fittings for Single-Use Systems. SterilEnz-II/AT connectors come with a platinum cured, medical grade silicone gasket mechanically attached to the fitting face. SterilEnz-II/AT fittings are made rugged inert polypropylene suitable for either autoclaving or gamma-irradiation. The new II/AT series is available in seven fitting sizes.