Learn More
Sigma Aldrich Fine Chemicals Biosciences SterilewareTM sampling spa
Catalog No. 502900502
Encompass
One-piece spatula is molded from rigid white polystyrene and retains approximately 15cc of material. Shape allows easy transfer of sample to container. Use to sample directly from production line or containers including bags and drums. V-shaped spatula is perfect for sampling granulated or semi solid materialSterile and individually packagedOne time use averts cross contaminationSharp point will pierce most packaging
Catalog No. 50-290-0502 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BAH369300000100EA
One-piece spatula is molded from rigid white polystyrene and retains approximately 15cc of material. Shape allows easy transfer of sample to container. Use to sample directly from production line or containers including bags and drums. V-shaped spatula is perfect for sampling granulated or semi solid materialSterile and individually packagedOne time use averts cross contaminationSharp point will pierce most packaging