Learn More
Baker Company Inc SterilGARD e3 w/ stand
SDP
Catalog No. NC1992181
Encompass
With the optimum balance of performance and energy efficiency, our biological safety cabinets protect personnel, product and the environment, all while increasing lab productivity and user comfort. Each cabinet is backed by years of research, customer feedback, and subjected to rigorous safety testing prior to shipment.The SterilGARD e3 biological safety cabinet from Baker offers a revolutionary airflow management system with proven containment technology that saves energy, increases productivity and improves comfort.
Catalog No. NC1992181 Supplier Baker Company Inc Supplier No. SG604
Add to Cart
Add to Cart
Request a Quote
With the optimum balance of performance and energy efficiency, our biological safety cabinets protect personnel, product and the environment, all while increasing lab productivity and user comfort. Each cabinet is backed by years of research, customer feedback, and subjected to rigorous safety testing prior to shipment.The SterilGARD e3 biological safety cabinet from Baker offers a revolutionary airflow management system with proven containment technology that saves energy, increases productivity and improves comfort.