Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
Certificate of Analysis
- Transparent labels for easy viewing and identification of medium
- Labels are laminated acetate to resist alcohol or chlorine treatment for cleanroom use
- Prepared bottles for sterility testing and microbial limits
Wide-Mouth Bottles
- Openings are 48mm in diameter to accommodate bulky samples
Order Info
Supplied in packs of 10 bottles
Compliance
Conform to harmonized USP requirements.
Specifications
Specifications
| Broth Type | Fluid Thioglycollate Medium |
| Cap Type | Screw |
| Certifications/Compliance | USP |
| Color | Light Beige |
| Content And Storage | 2°C to 8°C |
| Country of Origin | United States |
| Form | Liquid |
| Format | Bottle |
| Manufacturing Origin | United States |
| Media Type | Fluid Thioglycollate Medium |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?