Learn More
Sigma Aldrich Fine Chemicals Biosciences Sterility Indicator (Steam Sterilization), 25 strips
Catalog No. NC1865828
Encompass
These indicators consist of Bacillus stearothermophilus (ATCC 7953) spores impregnated on paper strips, individually placed into envelopes. Number of spores present per strip: 1 million.
Catalog No. NC1865828 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 7404125TESTSF
These indicators consist of Bacillus stearothermophilus (ATCC 7953) spores impregnated on paper strips, individually placed into envelopes. Number of spores present per strip: 1 million.