Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Cleanroom Products Sterility Maintenance Cover Pack
SDP

Catalog No. 19999151
Change view
Click to view available options
Dimensions (L x W):
24 x 24 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-999-151 24 x 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-999-151 Supplier Keystone Cleanroom Products Supplier No. 2000CPMSDIRR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pre-Sterilized Sterility Maintenance Covers are used to cover wrapped items after sterilization to provide protection from environmental factors, which could compromise sterility.

  • Pre-Sterilized Sterility Maintenance Covers are used to cover wrapped items after sterilization to provide protection from environmental factors, which could compromise sterility.
  • Made of a transparent, strong, 4 mil thick low density polyethylene material.
  • Each package is labeled with an expiration date and Lot number for complete traceability.
  • Made of a transparent, strong, 4 mil thick low density polyethylene material
  • Gamma irradiated and Validated Sterile to a 10-6 SAL
  • Lot controlled
  • Ready to use
  • 10 Sterility Maintemance Bags Per Double Poly Bag
  • Minimize contamination risks
  • Simplifies conformance to SOP's

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Certificate of Sterility
Closure Type Heat Seal
Color Clear
Disposable Single-use
Material LDPE
Sterility Sterile
Latex Free Yes
Sterilization Method Gamma
Dimensions (L x W) 24 x 24 in.
Length (English) 24 in.
Width (English) 24 in.
Product Type Cover
Length (Metric) 66 cm
Width (Metric) 66 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.