Learn More
Fisherbrand™ Sterilization Box for SterilElite Autoclave
Catalog No. STE16521012 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
SterilElite Autoclave STE16
SterilElite Autoclave STE24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
STE16521012 SterilElite Autoclave STE16
STE2452017 SterilElite Autoclave STE24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. STE16521012
Add to Cart
Add to Cart
A protective container used for sterilizing, transporting, and storing instruments in a controlled environment.
  • Stainless steel construction for durability and corrosion resistance venting holes
  • Open during sterilization to ensure full steam penetration
  • Closed after cycle to maintain sterility during transport/storage
  • Equipped with handles for safe and easy carrying
  • Protects contents from post-sterilization contamination
Specifications
For Use With (Equipment) SterilElite Autoclave STE16
Product Type Sterilization Box
Features Enables Multi-level Loading (Stacking Trays Vertically), Ensures Optimal Steam Circulation Between Trays, Provides Stable Positioning During Sterilization Cycles, Compatible With Full Tray Sets (Includes Tray Holders/Spring Holders), Corrosion-resistant Construction (Stainless Steel)
Material Stainless Steel
Quantity 1 Ea.
Voltage 100/240 V