Description
- Stainless steel construction for durability and corrosion resistance venting holes
- Open during sterilization to ensure full steam penetration
- Closed after cycle to maintain sterility during transport/storage
- Equipped with handles for safe and easy carrying
- Protects contents from post-sterilization contamination
Specifications
Specifications
| For Use With (Equipment) | SterilElite Autoclave STE24 |
| Product Type | Sterilization Box |
| Weight (English) | 8 lb. |
| Material | Stainless Steel |
| Quantity | 1 Ea. |
| Sterilization Time | 20 min. |
| Voltage | 100/240 V |