Learn More
Surgical Design™ Sterilization Cassette Double Button Tray With Removable Holding Clamps
Catalog No. 21124736 Shop All Surgical Design Products
Change view
Click to view available options
Size:
10
14
30
40
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-124-736 40
21-124-733 10
21-124-734 14
21-124-735 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-124-736 Supplier Surgical Design™ Supplier No. DSCDBDD15
Add to Cart
Add to Cart
Sterilization cassette trays with removable holding clamps provide secure, customizable instrument organization for cleaning and sterilization.
  • Available Slot Options: 10, 14, 20, and 30
  • Equipped with removable holding clamps to customize instrument positioning
  • Double-button locking mechanism keeps instruments secure during sterilization
  • Silicone rails protect instruments from abrasion and misalignment
  • Perforated stainless-steel construction ensures excellent steam and chemical penetration
  • Ideal for organizing delicate surgical, dental, pathology, and autopsy/postmortem instruments
  • Prevents damage, bending, or displacement during cleaning and autoclave cycles
  • Compatible with steam autoclaves and ultrasonic washers
  • Corrosion-resistant stainless steel for long-term reuse
Specifications
Product Type Sterilization Cassette and Storage
Disposable Reusable
Size 40
Sterility Non-sterile