Learn More
Surgical Design™ Sterilization Cassette Double Button Tray
Catalog No. 21124737 Shop All Surgical Design Products
Change view
Click to view available options
Size:
5
7
10
15
20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-124-737 5
21-124-738 7
21-124-739 10
21-124-740 15
21-124-741 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21-124-737 Supplier Surgical Design™ Supplier No. DSCDBSD05
Only null left
Add to Cart
Add to Cart
Double-button sterilization cassette trays are designed to securely organize and protect instruments during cleaning, sterilization, and storage.
  • Available Slot Options: 5, 7, 10, 15, and 20
  • Double-button locking system keeps instruments secure throughout sterilization
  • Rigid stainless-steel frame with silicone rails to protect instrument tips and edges
  • Perforated design allows optimal steam or chemical penetration
  • Prevents instrument shifting, bending, or damage during processing
  • Easy to load, unload, and organize surgical, dental, or autopsy instruments
  • Suitable for clinics, surgical centers, dental offices, and postmortem/autopsy labs
  • Compatible with ultrasonic cleaners and standard autoclaves
  • Corrosion-resistant and built for long-term reusable performance
Specifications
Product Type Sterilization Cassette and Storage
Disposable Reusable
Size 5
Sterility Non-sterile