Learn More
Sklar Instruments Container, Sklar Sterilization Half Size Container, Sklar 11 3/4 in. x 11 in. x 6 in. Sklar Sterilization Half Size Container Non-Perforated Bottom, Black. EA.
SDP
Catalog No. 501183137 Shop All Sklar Instruments Products
Encompass_Preferred
Container, SklarLite Sterilization Half Size Container, Sklar, 11 3/4 in. x 11 in. x 6 in. SklarLite Sterilization Half Size Container Non-Perforated Bottom, Black. SklarLite Sterilization Containers are made from anodized aluminum that allow them to be durable but lighter than traditional containers, ideal for not only sterilization and storage, but also easy transport. Available in multiple sizes, with lids available in a variety of colors, containers are available in sets or individual components. Sklar also offers a wide range of accessories, including baskets, silicone mats, custom ID labels, and filters. This product is a half size container with a non-perforated bottom and a black lid.
Catalog No. 50-118-3137 Supplier Sklar Instruments Supplier No. 104606
Container, SklarLite Sterilization Half Size Container, Sklar, 11 3/4 in. x 11 in. x 6 in. SklarLite Sterilization Half Size Container Non-Perforated Bottom, Black. SklarLite Sterilization Containers are made from anodized aluminum that allow them to be durable but lighter than traditional containers, ideal for not only sterilization and storage, but also easy transport. Available in multiple sizes, with lids available in a variety of colors, containers are available in sets or individual components. Sklar also offers a wide range of accessories, including baskets, silicone mats, custom ID labels, and filters. This product is a half size container with a non-perforated bottom and a black lid.