Learn More
Fine Science Tools Plastic Sterilization Containers w/ Silicone Mat, 27 x 16 x 4cm
SDP
Catalog No. NC9361224
Encompass
Product: Plastic Sterilization Container with Silicone Mat - 27 x 16 x 4cm/ Dimensions: 27 x 16 x 4cm/ Alloy / Material: Plastic/ Autoclave Safe: Yes/ Unit: Each
Catalog No. NC9361224 Supplier Fine Science Tools Supplier No. 2081002
Product: Plastic Sterilization Container with Silicone Mat - 27 x 16 x 4cm/ Dimensions: 27 x 16 x 4cm/ Alloy / Material: Plastic/ Autoclave Safe: Yes/ Unit: Each