Learn More
Med Vet International Sterilization Indicator Tape, Steam Autoclave, 1/2" Wide, J1495B, Each
Catalog No. 5000583447
Encompass_Preferred
Use as indication of sterility when color change occurs on the tape. • Tape will adhere to all types of wraps • Latex and lead free • Can be written on • Beige color • 50m (54yd) Sterilization Indicator Tape, Steam Autoclave, 1/2" Wide
Catalog No. 50-005-83447 Supplier Med Vet International Supplier No. JOR1495B
Use as indication of sterility when color change occurs on the tape. • Tape will adhere to all types of wraps • Latex and lead free • Can be written on • Beige color • 50m (54yd) Sterilization Indicator Tape, Steam Autoclave, 1/2" Wide