Learn More
World Precision Instrument Sterilization Mesh Mini-Casette, 4x4x2cm
SDP
Catalog No. 502539292
Encompass
Mesh sterilization baskets are perfect for sterilizing, storing and organizing small components or instrument. These baskets are manufactured from 304 stainlesss steel. They are non-magnetic and electropolished. They are burr-free to protect the instrument wraps from tearing. The fine mesh has open squares of 1.6 mm, and the wire edge diameter is 0.7 mm. These baskets come with a wire locking clip. This style of sterilization baskets is available in three sizes: 4x4x2 cm, 8x8x3.4 cm, and 10.5x7x2.5 cm.
Catalog No. 50-253-9292 Supplier World Precision Instrument Supplier No. 504766
Mesh sterilization baskets are perfect for sterilizing, storing and organizing small components or instrument. These baskets are manufactured from 304 stainlesss steel. They are non-magnetic and electropolished. They are burr-free to protect the instrument wraps from tearing. The fine mesh has open squares of 1.6 mm, and the wire edge diameter is 0.7 mm. These baskets come with a wire locking clip. This style of sterilization baskets is available in three sizes: 4x4x2 cm, 8x8x3.4 cm, and 10.5x7x2.5 cm.