Learn More
Andwin Scientific STERILIZATION POUCH 18X22
SDP
Catalog No. NC9271653
Encompass_Preferred
The 18" x 22" Convertors sterilization pouch are ideal for fast and easy presentation of sterile items. Each pouch has a chemical process indicator that inform the user of the sterilization process used. The pouches can be heat-sealed.
This item is not returnable. View return policy
Catalog No. NC9271653 Supplier Andwin Scientific Supplier No. 11213044
The 18" x 22" Convertors sterilization pouch are ideal for fast and easy presentation of sterile items. Each pouch has a chemical process indicator that inform the user of the sterilization process used. The pouches can be heat-sealed.