Learn More
Med Vet International Sterilization Pouch, 7 3/4" x 13", J0480C, 200-pack
Catalog No. 5000582291
Encompass_Preferred
Pre-formed pouches with self-adhering adhesive tape features one blue-tinted side for visualization of contents, with other side paper-backed. Will work with either steam autoclave or ethylene oxide gas sterilization.
Catalog No. 50-005-82291 Supplier Med Vet International Supplier No. JOR480C
Pre-formed pouches with self-adhering adhesive tape features one blue-tinted side for visualization of contents, with other side paper-backed. Will work with either steam autoclave or ethylene oxide gas sterilization.