Learn More
Hanna Pharmaceutical Supply Co., Inc. Sterilization Pouch Sure-Check Self-Seal 7.5x13
Catalog No. NC0511734
Encompass_Preferred
Sure-Check™ Self-Seal Sterilization Pouch Built-in muli-paramter internal/external process indicators that change color when all three criteria for sterilization have been met time, temperature and pressure (steam). 200/BXManufactured from medical grade paper with extra-wide chevron to protect against instrument breaching, thumb-notch for easy opening, 6 corner tack seals to prevent curling and other potential contaminants from entering pouch upon opening and a perforated fold to help ensure the adhesive strip forms an airtight sealOnce sterilized contents will remain sterile for up to 3 years provided the integrity of the pouch has not been compromisedFor use with steam and EtO sterilization processes only Not made with natural rubber latex 200/BX
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC0511734 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. HFGSCL2
Sure-Check™ Self-Seal Sterilization Pouch Built-in muli-paramter internal/external process indicators that change color when all three criteria for sterilization have been met time, temperature and pressure (steam). 200/BXManufactured from medical grade paper with extra-wide chevron to protect against instrument breaching, thumb-notch for easy opening, 6 corner tack seals to prevent curling and other potential contaminants from entering pouch upon opening and a perforated fold to help ensure the adhesive strip forms an airtight sealOnce sterilized contents will remain sterile for up to 3 years provided the integrity of the pouch has not been compromisedFor use with steam and EtO sterilization processes only Not made with natural rubber latex 200/BX
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.