Learn More
Sklar Instruments Sterilization Pouch Rack, Sklar, 4 in. x 18 in., Stainless Steel. This pouch rack is 4 inches x 18 inches and provides approximately 10 slots. EA/1.
SDP
Catalog No. 501437134 Shop All Sklar Instruments Products
Encompass_Preferred
Sterilization Pouch Rack, Sklar, 4 in. x 18 in., Stainless Steel. Sklar's Sterilization rack has a coil design to separate and hold sterilization pouches used with steam sterilization, EO Gas, and Gas Plasma. This pouch rack is 4 inches x 18 inches and provides approximately 10 slots.
This item is not returnable. View return policy
Catalog No. 50-143-7134 Supplier Sklar Instruments Supplier No. 101790
Sterilization Pouch Rack, Sklar, 4 in. x 18 in., Stainless Steel. Sklar's Sterilization rack has a coil design to separate and hold sterilization pouches used with steam sterilization, EO Gas, and Gas Plasma. This pouch rack is 4 inches x 18 inches and provides approximately 10 slots.