Learn More
Electron Microscopy Sciences Sterilization Tray
SDP
Catalog No. 50949687
Encompass_Preferred
A unique tray that allows you to place up to three diamond knives at one time in the autoclave. Protects the knives from any movement or damage.
Catalog No. 50-949-687 Supplier Electron Microscopy Sciences Supplier No. 72034
A unique tray that allows you to place up to three diamond knives at one time in the autoclave. Protects the knives from any movement or damage.