Learn More
Sklar Instruments Sterilization Tray, Sklar, 20-3/4 in. x 6-3/8 in. x 6 in., Stainless Steel. The overall size of this 9 qt tray is 20-3/4 in x 6-3/8 in x 6 in. EA/1.
SDP
Catalog No. 501437120 Shop All Sklar Instruments Products
Encompass_Preferred
Sterilization Tray, Sklar, 20-3/4 in. x 6-3/8 in. x 6 in., Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this 9 quart tray is 20-3/4 inches x 6-3/8 inches x 6 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7120 Supplier Sklar Instruments Supplier No. 101752
Sterilization Tray, Sklar, 20-3/4 in. x 6-3/8 in. x 6 in., Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this 9 quart tray is 20-3/4 inches x 6-3/8 inches x 6 inches.