Learn More
Med Vet International Sterilization Tray Two Layer Tray, J0041T, 10in x 6in x 1.5in, Each
Catalog No. 5000584397
Encompass_Preferred
This two layer tray can hold any type of instrument. The rubber mat allows instruments to lay in any configuration. It is autoclavable and can be cold sterilized.
Catalog No. 50-005-84397 Supplier Med Vet International Supplier No. JOR41T
This two layer tray can hold any type of instrument. The rubber mat allows instruments to lay in any configuration. It is autoclavable and can be cold sterilized.