Learn More
Electron Microscopy Sciences Sterilization Tray with Silicone Mat & Lid
SDP
Catalog No. 5030641
Encompass_Preferred
Perforated sterilizing tray is made from high strength Ultem plastic. The tray has a silicone mat which provides flexibility in the placement of the instruments for sterilization. A unique "grid" system exists in the base, tray and lid, making it easy to install the mat required, protecting your instruments properly. The trays are stackable and they have integral feet in base and corner ridges on lids which allow for easier handling in the autoclave and for storage.
All bases have pebble-type surface to keep mats from sticking.
Size: 151 x 64 x 32 mm
Catalog No. 50-306-41 Supplier Electron Microscopy Sciences Supplier No. 6611401
Perforated sterilizing tray is made from high strength Ultem plastic. The tray has a silicone mat which provides flexibility in the placement of the instruments for sterilization. A unique "grid" system exists in the base, tray and lid, making it easy to install the mat required, protecting your instruments properly. The trays are stackable and they have integral feet in base and corner ridges on lids which allow for easier handling in the autoclave and for storage.
All bases have pebble-type surface to keep mats from sticking.
Size: 151 x 64 x 32 mm