Learn More
Med Vet International Sterilization Tubing, 4" x 100', J0481C, 10 Count
Catalog No. 5000581511
Encompass_Preferred
Sterilization Tubing for sterile packaging of instruments. Cut off appropriate length from 100' roll. Blue-tinted side for visualization, other side paper-backed. Will work for either ethylene oxide or steam autoclaving.
Catalog No. 50-005-81511 Supplier Med Vet International Supplier No. JOR481C
Sterilization Tubing for sterile packaging of instruments. Cut off appropriate length from 100' roll. Blue-tinted side for visualization, other side paper-backed. Will work for either ethylene oxide or steam autoclaving.