Learn More
Beckman Coulter 39mL, Sterile + Certified Free Round Bottom Polypropylene Tube, 25 x 89mm - 50Pk
Catalog No. NC1857318
Encompass
This round bottom tube is designed for use in Beckman Coulter instruments and rotors. Quick-Seal tubes provides a secondary BioSafe containment. It also allows you to seal with a simple, hand-held sealing tool without plugs or O-rings. Quick-Seal tube spacer required.
Catalog No. NC1857318 Supplier Beckman Coulter Supplier No. C14304
This round bottom tube is designed for use in Beckman Coulter instruments and rotors. Quick-Seal tubes provides a secondary BioSafe containment. It also allows you to seal with a simple, hand-held sealing tool without plugs or O-rings. Quick-Seal tube spacer required.