Learn More
AVANTOR MASTERFLEX (VWR LLC) Sterilized Sampling Bottle Assembly, 500 mL
Catalog No. NC0706990
Encompass
Sterilized Sampling Bottle Assembly, 500 mL, Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to processSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are pre-sterilized by gamma irradiation¿eliminates labor and validation costs associated with assembly and cleaningStores up to 500 mL
Catalog No. NC0706990 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9564404
Sterilized Sampling Bottle Assembly, 500 mL, Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to processSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are pre-sterilized by gamma irradiation¿eliminates labor and validation costs associated with assembly and cleaningStores up to 500 mL