Learn More
Fisherbrand™ Sterilized Softside Safety Goggles
Catalog No. 128940023 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type: Sterile Goggles
Catalog No.
12-894-0023 Product Type: Sterile Goggles
1 options
This item is not returnable. View return policy
Catalog No. 12-894-0023
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
These goggles are anti-fogging and optically correct for clear vision. Used for safety in cleanroom environments.
  • Sufficient internal space, wide field of vision and large body design can be worn together with myopia glasses.
  • It is more durable against impact.
  • Arc design, in line with the facial contour of human face, more close and comfortable fit with the face and nose wing.
  • Fully adjustable and comfortable headband, fits all head sizes.
  • Double side anti fog, Clearer sight, light transmittance is over 90%.
  • Softside pvc construction that delivers a comfortable fitting and allows for good peripheral vision.
  • Ethylene Oxide (EtO) Sterilized.
Specifications
Lens Tint Clear
Product Type Sterile Goggles
Dimensions (L x W x H) 190.71 x 70.32 x 73.32 mm
Magnification Power Transmittance
Material PVC
Disposable Single-use
Lens Material PVC
Frame Material PVC