Learn More
CROWN FOOD SERVICE EQUIPMENT Market Forge Sterilmatic Sterilizer Autoclave, Analog, Fixed Temp At 250F, 208V/240V 60Hz
Catalog No. NC2180814
Encompass_Preferred
Sterilize Glass Beakers, Pipettes, Test Tubes And Flasks. Metal And Plastic Instruments, Syringes, Needles, Brushes, Rubber Gloves, Catheters, Rubber Tubing, Dressings, Linens, Petri Dishes And More. Standard Features: Automatic Temperature ControlsTemperature, Fixed At 250°F (121°C)Thermostatic Steam TrapThermometer Range 180°-300°F (82°-149°C)0-30 Lb. Steam Gauge0-60 Minute TimerLow Water Cut-Off, Signal Light, And Safety ValveFlat Perforated Steam BaffleRemovable Pan Supports
This item is not returnable. View return policy
Catalog No. NC2180814 Supplier CROWN FOOD SERVICE EQUIPMENT Supplier No. STME
Add to Cart
Add to Cart
Request a Quote
Sterilize Glass Beakers, Pipettes, Test Tubes And Flasks. Metal And Plastic Instruments, Syringes, Needles, Brushes, Rubber Gloves, Catheters, Rubber Tubing, Dressings, Linens, Petri Dishes And More. Standard Features: Automatic Temperature ControlsTemperature, Fixed At 250°F (121°C)Thermostatic Steam TrapThermometer Range 180°-300°F (82°-149°C)0-30 Lb. Steam Gauge0-60 Minute TimerLow Water Cut-Off, Signal Light, And Safety ValveFlat Perforated Steam BaffleRemovable Pan Supports