Learn More
Fine Science Tools Hot Bead Sterilizers, 14.5 x 12.5 x 13.5 cm
SDP
Supplier:  Fine Science Tools 1800045
Encompass
Product: Hot Bead Sterilizer - 8cm Well/ Dimensions: 14.5 x 12.5 x 13.5cm/ Well Depth: 8cm/ Well Diameter: 4cm/ Well Volume: 100cm3 Unit: Each
Catalog No. NC9477349