Learn More
Research Products International Corp Sterilizing Benchtop Incubator
SDP
Catalog No. 501976847
Encompass
This unique incubator features a built in overhead 254nm UV lamp that sterilizes the chamber between critical experiments. UV and temperature functions operate independently from each other. Ideal for molecular biology applications such as growth promotion studies, phage and cosmid cloning and BL3 laboratory operations. Forced draft incubation technology using precise microprocessor controls maintains constant and uniform air circulation with minimal airflow distrubance.
Additional features include: automatic UV shutoff when door is open; stainless steel interior; two stainless steel adjustable shelves; clear acrylic door with gasket seal and compact space saving design. Supplied complete with two shelves, 254nm UV lamp and instructions.
Twelve month warranty.
Catalog No. 50-197-6847 Supplier Research Products International Corp Supplier No. 95035801
This unique incubator features a built in overhead 254nm UV lamp that sterilizes the chamber between critical experiments. UV and temperature functions operate independently from each other. Ideal for molecular biology applications such as growth promotion studies, phage and cosmid cloning and BL3 laboratory operations. Forced draft incubation technology using precise microprocessor controls maintains constant and uniform air circulation with minimal airflow distrubance.
Additional features include: automatic UV shutoff when door is open; stainless steel interior; two stainless steel adjustable shelves; clear acrylic door with gasket seal and compact space saving design. Supplied complete with two shelves, 254nm UV lamp and instructions.
Twelve month warranty.