Learn More
Sigma Aldrich Fine Chemicals Biosciences Sterilizing pans capacity1EA
Catalog No. 502901184
Encompass
Autoclavable natural polypropylene with rolled edges for stiffness and handles for carrying ease. Nestable when empty.
Catalog No. 50-290-1184 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3603681EA
Autoclavable natural polypropylene with rolled edges for stiffness and handles for carrying ease. Nestable when empty.