Learn More
Research Products International Corp Sterilizing Tray, 19 x 10 1/2 x 5 1/8 Inches
SDP
Catalog No. 50212955
Encompass
Made of heavy duty polypropylene that is steam autoclavable to 250°F (121°C). Feature smooth surfaces and rounded corners for ease of cleaning. Trays are conveniently tapered for nesting during storage.
Catalog No. 50-212-955 Supplier Research Products International Corp Supplier No. H16262
Made of heavy duty polypropylene that is steam autoclavable to 250°F (121°C). Feature smooth surfaces and rounded corners for ease of cleaning. Trays are conveniently tapered for nesting during storage.