Learn More
Research Products International Corp Sterilizing Tray, 12 x 7 3/4 x 5 1/8 Inches
SDP
Catalog No. 50212953
Encompass
Made of heavy duty polypropylene that is steam autoclavable to 250°F (121°C). Feature smooth surfaces and rounded corners for ease of cleaning. Trays are conveniently tapered for nesting during storage.
Catalog No. 50-212-953 Supplier Research Products International Corp Supplier No. H16260
Made of heavy duty polypropylene that is steam autoclavable to 250°F (121°C). Feature smooth surfaces and rounded corners for ease of cleaning. Trays are conveniently tapered for nesting during storage.