Learn More
World Precision Instrument Instrument Sterilization Tray w Silicone Pads, 23x15x5cm
SDP
Catalog No. NC1943204
Encompass
The unique design allows for better steam circulation and faster drying. If sterilization is imperative, use sterilization wraps, heat-sealed plastic and paper pouches (woven and non-woven) before placing trays in an autoclave. The packaging material must allow sterilant penetration, provide protection against contact contamination during handling, provide an effective barrier to microbial penetration, and keep the processed item sterile after sterilization.
Catalog No. NC1943204 Supplier World Precision Instrument Supplier No. 501934
The unique design allows for better steam circulation and faster drying. If sterilization is imperative, use sterilization wraps, heat-sealed plastic and paper pouches (woven and non-woven) before placing trays in an autoclave. The packaging material must allow sterilant penetration, provide protection against contact contamination during handling, provide an effective barrier to microbial penetration, and keep the processed item sterile after sterilization.