Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Coverage™ Spray HB Plus Ready-to-Use Disinfectant Cleaner

Catalog No. 1441519 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
22 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
14-415-19 22 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-415-19 Supplier STERIS Supplier No. 162477

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quaternary-based, one-step disinfectant for hard surfaces

  • Provides a broad range of activity against gram-positive and gram-negative bacteria, viruses, and fungi
  • Effective in 5% organic soil

Compliance

Complies with OSHA Bloodborne Pathogen Standard (29 CFR Part 1910.1030).

Specifications

Form Liquid
pH 11
Appearance Clear
Certifications/Compliance OSHA
Composition 0.05% Di-n-alkyl Dimethyl Ammonium Chloride, 0.03% N-alkyl Dimethyl Benzyl Ammonium Chloride, 0.017%
Container Type Trigger spray bottle
For Use With (Application) Used as Bactericidal, Fungicidal and Virucidal in the presence of 5% Organic Serum against Bacteria, Pseudomonas, Staph, Salmonella, VRE, E coli, HBV and the AIDS Virus
Volume (English) 22 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.