Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS LpH™ se One-Step Germicidal Detergent

Catalog No. 1441510 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
14-415-10 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-415-10 Supplier STERIS Supplier No. 646608
This item is not returnable. View return policy

One-step nonalkaline disinfectant cleaner and deodorizer for use on hard, nonporous surfaces

Ideal for use in a rotation program with Vesphene™ IIse Disinfectant (14-415-11).

  • Phenolic-based germicidal detergent
  • Kills a broad spectrum of common bacteria at a use dilution rate of 1:256 without harming surfaces
  • Also tuberculocidal and HIV-effective
  • Effective with soft water or hard water up to 400ppm hardness (as CaCO3) and in the presence of 5% serum

Compliance

Meets OSHA guidelines for cleaning and disinfecting blood spills.

Specifications

Form Liquid
pH 2.6
Appearance Yellow to Amber
Certifications/Compliance NOISH, OSHA
Composition 5 to 10% p-tertiary Amylphenol, 5 to 10% Phenyl phenol, 15% Phosphoric Acid, 7.5% Isopropanol, <5
For Use With (Application) Kills a broad spectrum of common Bacteria on hard, non porous environmental surfaces such as floors, walls, counters, tools, carts and other equipment in hospitals, nursing homes, clinics, pharmaceutical manufacturing facilities and many more
Volume (English) 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.