Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Staphene™ Spray Environmental Disinfectant

Catalog No. 1441515 Shop All STERIS Corporation Products
Change view
Click to view available options
Staphene Spray; 12 x 16 net Wt./Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
14-415-15 Staphene Spray; 12 x 16 net Wt./Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-415-15 Supplier STERIS Supplier No. 6389Y3
This item is not returnable. View return policy

Ready-to-use, aerosol disinfectant and deodorizer for hard surfaces

  • Alcohol-based solution of phenolics, quaternary ammonium compounds, and deodorizing compounds
  • Effective against a broad range of organisms, including Mycobacterium tuberculosis var. bovis and HIV-1 virus
  • Bactericidal, fungicidal, and virucidal in 5% serum

Compliance

Satisfies the OSHA Bloodborne Pathogen Guidelines (29 CFR Part 1910.1030) for cleaning blood and body fluid spills.

Specifications

Form Liquid
Appearance Clear
Certifications/Compliance OSHA
Composition 0.077% o-Benzyl-P-Chlorophenol, 0.074% p-tertiary Amylphenol, 0.041% 2-Phenylphenol, 0.042% n-Alkyl
Content And Storage <100°F
For Use With (Application) Used in medical and dental offices, sick rooms, nursing homes and hospitals for non porous environmental surfaces with broad biocidal action for cleaning Blood and body fluid spills
Volume (English) 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.